accessibility menu, dialog, popup

UserName

CAS RN 55-91-4

IUPAC Name: bis(propan-2-yl) phosphorofluoridate
Synonyms: isoflurophate isoflurophate (substance) fluophosphoric acid, diisopropyl ester rcra waste number P043 diflurophate fluoropryl diisopropyl phosphofluoridate floropryl isoflurophate (product) diisopropyl ester fluophosphoric acid
Molecular Weight (g/mol): 184.15
Molecular Formula: C6H14FO3P
InChi Key: MUCZHBLJLSDCSD-UHFFFAOYSA-N
SMILES: CC(C)OP(F)(=O)OC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)