CAS RN 5497-67-6
IUPAC Name:
2,2-dimethylpent-4-enal
Synonyms:
2,2-dimethylpent-4-enal
Molecular Weight (g/mol):
112.17
Molecular Formula:
C7H12O
InChi Key:
DXSDIWHOOOBQTJ-UHFFFAOYSA-N
SMILES:
CC(C)(CC=C)C=O
1
–
1
of
1
results
1
Promotions Available
2,2-Dimethyl-4-pentenal, 97%, stabilized
2,2-Dimethyl-4-pentenal, 97%, stabilized
CAS: 5497-67-6 Molecular Formula: C7H12O Molecular Weight (g/mol): 112.17 MDL Number: MFCD00010699 InChI Key: DXSDIWHOOOBQTJ-UHFFFAOYSA-N Synonym: 2,2-dimethyl-4-pentenal,4-pentenal, 2,2-dimethyl,2,2-dimethyl-pent-4-enal,4-pentenal,2,2-dimethyl,zlchem 644,acmc-20aojn,pubchem22726,2,2-dimethyl4-pentenal,4-pentenal,2-dimethyl,2,2-dimethy-4-pentenal PubChem CID: 79636 IUPAC Name: 2,2-dimethylpent-4-enal SMILES: CC(C)(CC=C)C=O