accessibility menu, dialog, popup

UserName

NHNCl

CAS RN 5496-32-2

IUPAC Name: 2-(4-chlorophenyl)-4,5-diphenyl-1H-imidazole
Synonyms: 2-(4-chlorophenyl)-4,5-diphenyl-1H-imidazole
Molecular Weight (g/mol): 330.82
Molecular Formula: C21H15ClN2
InChi Key: NJXBHEMXPUMJRO-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results