accessibility menu, dialog, popup

UserName

H3C(R)CH3OO

CAS RN 54947-74-9

IUPAC Name: (4R)-4-methyloctanoate
Synonyms: (4R)-4-methyloctanoate
Molecular Weight (g/mol): 157.23
Molecular Formula: C9H17O2
InChi Key: LEGGANXCVQPIAI-MRVPVSSYSA-M
SMILES: CCCC[C@@H](C)CCC([O-])=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)