accessibility menu, dialog, popup

UserName

CAS RN 54903-10-5

ClOHNOO

CAS RN 54903-10-5

IUPAC Name: 6-(2-chloroacetyl)-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 6-(2-chloroacetyl)-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol): 211.6
Molecular Formula: C9H6ClNO3
InChi Key: KOOWFDRPKXUCRF-UHFFFAOYSA-N
SMILES: ClCC(=O)C1=CC=C2NC(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)