accessibility menu, dialog, popup

UserName

CAS RN 54812-86-1

H3COHCH3HS

CAS RN 54812-86-1

IUPAC Name: 3-sulfanylbutan-2-ol
Synonyms: 3-sulfanylbutan-2-ol
Molecular Weight (g/mol): 106.18
Molecular Formula: C4H10OS
InChi Key: MJQWABQELVFQJL-UHFFFAOYNA-N
SMILES: CC(O)C(C)S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results