accessibility menu, dialog, popup

UserName

NaNaOHNHNNOOOSOOOSOOO(E)

CAS RN 548-80-1

IUPAC Name: disodium (3E)-5-hydroxy-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (3E)-5-hydroxy-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 513.36
Molecular Formula: C16H9N3Na2O10S2
InChi Key: MQHFURHHKNKIRN-GVLVODDDSA-L
SMILES: [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)