accessibility menu, dialog, popup

UserName

ON

CAS RN 5470-96-2

IUPAC Name: quinoline-2-carbaldehyde
Synonyms: 2-quinolinecarboxaldehyde
Molecular Weight (g/mol): 157.17
Molecular Formula: C10H7NO
InChi Key: WPYJKGWLDJECQD-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2C=CC=CC2=N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)