CAS RN 545375-30-2
IUPAC Name:
2-aminoethyl benzoate hydrochloride
Synonyms:
2-aminoethyl benzoate hydrochloride
Molecular Weight (g/mol):
201.65
Molecular Formula:
C9H12ClNO2
InChi Key:
ZXTMLOSVFWKYCL-UHFFFAOYSA-N
SMILES:
Cl.NCCOC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals
Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals
CAS: 545375-30-2 Molecular Formula: C9H12ClNO2 Molecular Weight (g/mol): 201.65 MDL Number: MFCD09953460 InChI Key: ZXTMLOSVFWKYCL-UHFFFAOYSA-N Synonym: ethanolamine benzoate hydrochloride,2-aminoethyl benzoate hydrochloride,ethanolamine benzoate hydrochloride, 98+%,2-benzoyloxyethylamine hydrochloride PubChem CID: 67778770 IUPAC Name: 2-aminoethyl benzoate;hydrochloride SMILES: Cl.NCCOC(=O)C1=CC=CC=C1