CAS RN 544-00-3
CAS RN 544-00-3
IUPAC Name:
bis(3-methylbutyl)amine
Synonyms:
bis(3-methylbutyl)amine
Molecular Weight (g/mol):
157.3
Molecular Formula:
C10H23N
InChi Key:
SPVVMXMTSODFPU-UHFFFAOYSA-N
SMILES:
CC(C)CCNCCC(C)C
1
–
1
of
1
results
1
Diisoamylamine 97.0+%, TCI America™
CAS: 544-00-3 Molecular Formula: C10H23N Molecular Weight (g/mol): 157.301 MDL Number: MFCD00015053 InChI Key: SPVVMXMTSODFPU-UHFFFAOYSA-N Synonym: diisopentylamine,diisoamylamine,1-butanamine, 3-methyl-n-3-methylbutyl,di 3-methylbutyl amine,bis 3-methylbutyl amine,di-iso-pentylamine = di-iso-amylamine,3-methyl-n-3-methylbutyl butan-1-amine,diisoamylamin,di-iso-amylamine,acmc-1astz PubChem CID: 10988 IUPAC Name: 3-methyl-N-(3-methylbutyl)butan-1-amine SMILES: CC(C)CCNCCC(C)C