CAS RN 54394-90-0
IUPAC Name:
disodium hydrogen ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms:
disodium hydrogen {[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol):
447.14
Molecular Formula:
C9H13N3Na2O11P2
InChi Key:
ZGDYAAWYYNVXEB-WFIJOQBCSA-L
SMILES:
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]1O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Cytidine-5'-diphosphate disodium salt, 95%
CAS: 54394-90-0 Molecular Formula: C9H13N3Na2O11P2 Molecular Weight (g/mol): 447.14 MDL Number: MFCD00084682 InChI Key: ZGDYAAWYYNVXEB-WFIJOQBCSA-L Synonym: CDP disodium salt PubChem CID: 131632379 IUPAC Name: disodium;(2R,3R,4S,5S)-2-(4-amino-2-oxopyrimidin-1-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate SMILES: [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]1O