accessibility menu, dialog, popup

UserName

CAS RN 5435-64-3

H3COCH3CH3H3C

CAS RN 5435-64-3

IUPAC Name: 3,5,5-trimethylhexanal
Synonyms: trimethylhexanal
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: WTPYRCJDOZVZON-UHFFFAOYNA-N
SMILES: CC(CC=O)CC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results