accessibility menu, dialog, popup

UserName

CAS RN 5432-85-9

IUPAC Name: 4-(propan-2-yl)cyclohexan-1-one
Synonyms: 4-isopropylcyclohexan-1-one
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: FPKISACHVIIMRA-UHFFFAOYSA-N
SMILES: CC(C)C1CCC(=O)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)