accessibility menu, dialog, popup

UserName

ONONCl

CAS RN 54231-32-2

IUPAC Name: 3-chloro-2-nitropyridine
Synonyms: 3-chloro-2-nitropyridine
Molecular Weight (g/mol): 158.54
Molecular Formula: C5H3ClN2O2
InChi Key: PSGASDJUCYTRAD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=NC=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results