accessibility menu, dialog, popup

UserName

CAS RN 54078-29-4

IUPAC Name: 6,10-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-8-one
Synonyms: 1,8-diazafluoren-9-one
Molecular Weight (g/mol): 182.18
Molecular Formula: C11H6N2O
InChi Key: FOSUVSBKUIWVKI-UHFFFAOYSA-N
SMILES: O=C1C2=C(C=CC=N2)C2=C1N=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results