accessibility menu, dialog, popup

UserName

CAS RN 5407-98-7

O

CAS RN 5407-98-7

IUPAC Name: cyclobutyl(phenyl)methanone
Synonyms: cyclobutyl-phenyl-methanone
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: MVEBDOSCXOQNAR-UHFFFAOYSA-N
SMILES: O=C(C1CCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results