accessibility menu, dialog, popup

UserName

CAS RN 5407-91-0

O

CAS RN 5407-91-0

IUPAC Name: 1,4-diphenylbutan-1-one
Synonyms: 1,4-diphenylbutan-1-one
Molecular Weight (g/mol): 224.3
Molecular Formula: C16H16O
InChi Key: GBUMEGLMTNAXOM-UHFFFAOYSA-N
SMILES: O=C(CCCC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results