CAS RN 5405-79-8
IUPAC Name:
2,2-dimethylhexan-3-one
Synonyms:
3-hexanone, 2,2-dimethyl-
Molecular Weight (g/mol):
128.22
Molecular Formula:
C8H16O
InChi Key:
PYCHXHVFOZBVEY-UHFFFAOYSA-N
SMILES:
CCCC(=O)C(C)(C)C
1
–
1
of
1
results
1
Promotions Available
2,2-Dimethyl-3-hexanone, 99%
2,2-Dimethyl-3-hexanone, 99%
CAS: 5405-79-8 Molecular Formula: C8H16O Molecular Weight (g/mol): 128.22 MDL Number: MFCD00027140 InChI Key: PYCHXHVFOZBVEY-UHFFFAOYSA-N Synonym: 2,2-dimethyl-3-hexanone,3-hexanone, 2,2-dimethyl,tert-butyl propyl ketone,acmc-20aojp,3-hexanone,2-dimethyl,2,2-dimethylhexane-3-one,2,2-dimethyl-hexan-3-one,3-hexanone,2,2-dimethyl PubChem CID: 221168 IUPAC Name: 2,2-dimethylhexan-3-one SMILES: CCCC(=O)C(C)(C)C