accessibility menu, dialog, popup

UserName

FFFNHOHNO

CAS RN 54-20-6

IUPAC Name: 5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: trifluoromethyluracil
Molecular Weight (g/mol): 180.09
Molecular Formula: C5H3F3N2O2
InChi Key: LMNPKIOZMGYQIU-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CNC(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results