accessibility menu, dialog, popup

UserName

CAS RN 53934-76-2

IUPAC Name: 2-(2-oxopyrrolidin-1-yl)acetate
Synonyms: 2-(2-oxopyrrolidin-1-yl)acetate
Molecular Weight (g/mol): 142.14
Molecular Formula: C6H8NO3
InChi Key: JGPIWNNFLKDTSR-UHFFFAOYSA-M
SMILES: [O-]C(=O)CN1CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results