accessibility menu, dialog, popup

UserName

NaNaPdClClClClClCl

CAS RN 53823-60-2

IUPAC Name: disodium hexachloropalladiumbis(ylium)
Synonyms: disodium hexachloropalladiumbis(ylium)
Molecular Weight (g/mol): 365.1
Molecular Formula: Cl6Na2Pd
InChi Key: OWJQEDIJRBKDJO-UHFFFAOYSA-H
SMILES: [Na+].[Na+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results