CAS RN 53721-12-3
IUPAC Name:
1,1'-diethyl-[4,4'-bipyridine]-1,1'-diium dibromide
Synonyms:
1,1'-diethyl-[4,4'-bipyridine]-1,1'-diium dibromide
Molecular Weight (g/mol):
374.12
Molecular Formula:
C14H18Br2N2
InChi Key:
LCEBDKLPALDQPV-UHFFFAOYSA-L
SMILES:
[Br-].[Br-].CC[N+]1=CC=C(C=C1)C1=CC=[N+](CC)C=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl viologen dibromide, 97%, Thermo Scientific™
Ethyl viologen dibromide, 97%, Thermo Scientific™
CAS: 53721-12-3 Molecular Formula: C14H18Br2N2 Molecular Weight (g/mol): 374.11 InChI Key: LCEBDKLPALDQPV-UHFFFAOYSA-L Synonym: ethyl viologen dibromide,1,1'-diethyl-4,4'-bipyridinium dibromide,4,4'-bipyridinium, 1,1'-diethyl-, dibromide,ethylviologen dibromide,1,1'-diethyl-4,4'-bipyridiniumdibromide,1,1/'-diethyl-4,4/'-bipyridinium dibromide,1,1-diethyl-4,4-bipyridinium dibromide,1,1'-diethyl-4,4'-bipyridine-1,1'-diium bromide,1-ethyl-4-1-ethyl 4-pyridyl pyridine, bromide, bromide PubChem CID: 11474262 IUPAC Name: 1-ethyl-4-(1-ethylpyridin-1-ium-4-yl)pyridin-1-ium;dibromide SMILES: CC[N+]1=CC=C(C=C1)C2=CC=[N+](C=C2)CC.[Br-].[Br-]