accessibility menu, dialog, popup

UserName

HClHN

CAS RN 537-67-7

IUPAC Name: hydrogen N-phenylaniline chloride
Synonyms: hydrogen diphenylamine chloride
Molecular Weight (g/mol): 205.69
Molecular Formula: C12H12ClN
InChi Key: JEFJSEIUEJBMSR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].N(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results