CAS RN 53595-65-6
CAS RN 53595-65-6
IUPAC Name:
5-bromothiophene-2-sulfonamide
Synonyms:
5-bromothiophene-2-sulfonamide
Molecular Weight (g/mol):
242.11
Molecular Formula:
C4H4BrNO2S2
InChi Key:
WXJQQLDICAOBJB-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=CC=C(Br)S1
1
–
1
of
1
results
1
5-Bromo-2-thiophenesulfonamide 97.0+%, TCI America™
CAS: 53595-65-6 Molecular Formula: C4H4BrNO2S2 Molecular Weight (g/mol): 242.11 MDL Number: MFCD00067990 InChI Key: WXJQQLDICAOBJB-UHFFFAOYSA-N Synonym: 5-bromo-2-thiophenesulfonamide,5-bromo-thiophene-2-sulfonic acid amide,2-thiophenesulfonamide, 5-bromo,5-bromothiophene-2-sulfonic acid amide,5-bromothiophene-2-sulphonamide,2-bromo thiophene-5-sulfonamide,5-bromothiophene-2-sulfonylamine,pubchem7369,maybridge1_004885,acmc-1b1sa PubChem CID: 1241304 IUPAC Name: 5-bromothiophene-2-sulfonamide SMILES: NS(=O)(=O)C1=CC=C(Br)S1