accessibility menu, dialog, popup

UserName

OONSN

CAS RN 53572-98-8

IUPAC Name: imidazo[2,1-b][1,3]thiazole-6-carboxylate
Synonyms: imidazo[2,1-b][1,3]thiazole-6-carboxylate
Molecular Weight (g/mol): 167.16
Molecular Formula: C6H3N2O2S
InChi Key: BAKIBSRDBASFAQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CN2C=CSC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)