CAS RN 5348-51-6
IUPAC Name:
hydrogen 6-methyl-1,2-dihydropyrimidin-2-one chloride
Synonyms:
hydrogen 2-pyrimidone,6-methyl chloride
Molecular Weight (g/mol):
146.57
Molecular Formula:
C5H7ClN2O
InChi Key:
FDRYQDWXFZBBAD-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=CC=NC(=O)N1
1
–
1
of
1
results
1
2-Hydroxy-4-methylpyrimidine Hydrochloride 98.0+%, TCI America™
CAS: 5348-51-6 Molecular Formula: C5H7ClN2O Molecular Weight (g/mol): 146.574 MDL Number: MFCD00151347 InChI Key: FDRYQDWXFZBBAD-UHFFFAOYSA-N Synonym: 4-methylpyrimidin-2-ol hydrochloride,2-hydroxy-4-methylpyrimidine hydrochloride,2-hydroxy-4-methylpyrimidine hcl,6-methylpyrimidin-2 1h-one hydrochloride,2-hydroxy-4-methylpyrimidine hydro-chloride,2 1h-pyrimidinone, 4-methyl-, monohydrochloride,4-methyl-3-hydropyrimidin-2-one, chloride,2-hydroxy-4-methylpyrimidinehydrochloride,zlchem 746,pubchem22005 PubChem CID: 2723648 IUPAC Name: 6-methyl-1H-pyrimidin-2-one;hydrochloride SMILES: CC1=CC=NC(=O)N1.Cl