CAS RN 532967-21-8
IUPAC Name:
2,6-difluoro-4-hydroxybenzaldehyde
Synonyms:
2,6-difluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol):
158.1
Molecular Formula:
C7H4F2O2
InChi Key:
ROAQMGJHSNIROA-UHFFFAOYSA-N
SMILES:
OC1=CC(F)=C(C=O)C(F)=C1
1
–
1
of
1
results
1
Promotions Available
2,6-Difluoro-4-hydroxybenzaldehyde, 95%
2,6-Difluoro-4-hydroxybenzaldehyde, 95%
CAS: 532967-21-8 Molecular Formula: C7H4F2O2 Molecular Weight (g/mol): 158.104 MDL Number: MFCD07772042 InChI Key: ROAQMGJHSNIROA-UHFFFAOYSA-N Synonym: 2,6-difluoro-4-hydroxy-benzaldehyde,benzaldehyde, 2,6-difluoro-4-hydroxy,ksc495q9f,3,5-difluoro-4-formylphenol,4-hydroxy-2,6-difluorobenzaldehyde,2,6-difluoro-4-hydroxy benzaldehyde,2,6-bis fluoranyl-4-oxidanyl-benzaldehyde,benzaldehyde, 2,6-difluoro-4-hydroxy-9ci PubChem CID: 24903548 IUPAC Name: 2,6-difluoro-4-hydroxybenzaldehyde SMILES: C1=C(C=C(C(=C1F)C=O)F)O