accessibility menu, dialog, popup

UserName

CAS RN 5323-47-7

H3CNCH3ONOO

CAS RN 5323-47-7

IUPAC Name: N,N-diethyl-4-nitrobenzamide
Synonyms: N,N-diethyl-4-nitrobenzamide
Molecular Weight (g/mol): 222.24
Molecular Formula: C11H14N2O3
InChi Key: JLZWKZDHHREXTA-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results