accessibility menu, dialog, popup

UserName

HClNH2H2NNOO

CAS RN 53209-19-1

IUPAC Name: hydrogen 4-nitrobenzene-1,2-diamine chloride
Synonyms: hydrogen 2-amino-4-nitroaniline chloride
Molecular Weight (g/mol): 189.6
Molecular Formula: C6H8ClN3O2
InChi Key: IFTLGFVLGVXRBT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(C=C1N)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results