accessibility menu, dialog, popup

UserName

HOOHOOOHOOHO

CAS RN 53159-92-5

IUPAC Name: cyclobutane-1,2,3,4-tetracarboxylic acid
Synonyms: cyclobutane-1,2,3,4-tetracarboxylic acid
Molecular Weight (g/mol): 232.14
Molecular Formula: C8H8O8
InChi Key: CURBACXRQKTCKZ-UHFFFAOYSA-N
SMILES: OC(=O)C1C(C(C1C(O)=O)C(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results