accessibility menu, dialog, popup

UserName

CH3CH3NNHN

CAS RN 53112-28-0

IUPAC Name: 4,6-dimethyl-N-phenylpyrimidin-2-amine
Synonyms: pyrimethanil
Molecular Weight (g/mol): 199.26
Molecular Formula: C12H13N3
InChi Key: ZLIBICFPKPWGIZ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(NC2=CC=CC=C2)=N1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results