accessibility menu, dialog, popup

UserName

HOOPOOO

CAS RN 5301-78-0

IUPAC Name: 4-(hydroxymethyl)-2,6,7-trioxa-1λ⁵-phosphabicyclo[2.2.2]octan-1-one
Synonyms: 4-(hydroxymethyl)-2,6,7-trioxa-1λ⁵-phosphabicyclo[2.2.2]octan-1-one
Molecular Weight (g/mol): 180.1
Molecular Formula: C5H9O5P
InChi Key: YASRHLDAFCMIPB-UHFFFAOYSA-N
SMILES: OCC12COP(=O)(OC1)OC2
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)