CAS RN 52787-14-1
IUPAC Name:
methyl 4-(2-methoxy-2-oxoethyl)benzoate
Synonyms:
methyl 4-(2-methoxy-2-oxoethyl)benzoate
Molecular Weight (g/mol):
208.21
Molecular Formula:
C11H12O4
InChi Key:
QAQYBHOZQQRJBA-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=CC=C(C=C1)C(=O)OC
1
–
1
of
1
results
1
Dimethyl Homoterephthalate 98.0+%, TCI America™
CAS: 52787-14-1 Molecular Formula: C11H12O4 Molecular Weight (g/mol): 208.213 MDL Number: MFCD07779210 InChI Key: QAQYBHOZQQRJBA-UHFFFAOYSA-N Synonym: Homoterephthalic Acid Dimethyl Ester, Methyl 4-Methoxycarbonylphenylacetate, 4-Methoxycarbonylphenylacetic Acid Methyl Ester, Methyl 4-(2-Methoxy-2-oxoethyl)benzoate, 4-(2-Methoxy-2-oxoethyl)benzoic Acid Methyl Ester PubChem CID: 10536335 IUPAC Name: methyl 4-(2-methoxy-2-oxoethyl)benzoate SMILES: COC(=O)CC1=CC=C(C=C1)C(=O)OC