accessibility menu, dialog, popup

UserName

HOONHOOHO

CAS RN 5262-39-5

IUPAC Name: 2-(carboxyformamido)acetic acid
Synonyms: N-oxalylglycine
Molecular Weight (g/mol): 147.09
Molecular Formula: C4H5NO5
InChi Key: BIMZLRFONYSTPT-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results