accessibility menu, dialog, popup

UserName

H3COOHO

CAS RN 52490-15-0

IUPAC Name: 4-(2-butyl-1-benzofuran-3-carbonyl)phenol
Synonyms: 4-(2-butyl-1-benzofuran-3-carbonyl)phenol
Molecular Weight (g/mol): 294.35
Molecular Formula: C19H18O3
InChi Key: ZHGKQUXXASLVQQ-UHFFFAOYSA-N
SMILES: CCCCC1=C(C(=O)C2=CC=C(O)C=C2)C2=CC=CC=C2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results