accessibility menu, dialog, popup

UserName

CAS RN 52232-67-4

IUPAC Name: (4S)-4-{[(1S,2S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-[({[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-3-carbamoyl-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-2-carbamoyl-1-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}ethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-2-carboxyethyl]carbamoyl}propyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}pentyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-3-methylbutyl]carbamoyl}-2-(1H-indol-3-yl)ethyl]carbamoyl}-3-carboxypropyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-3-carboxypropyl]carbamoyl}-3-(methylsulfanyl)propyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-2-carbamoylethyl]carbamoyl}-3-methylbutyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}pentyl]carbamoyl}methyl)carbamoyl]-3-methylbutyl]carbamoyl}-2-carbamoylethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}-3-(methylsulfanyl)propyl]carbamoyl}-3-methylbutyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-2-methylbutyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-amino-3-hydroxypropanamido]-3-methylbutanamido]-3-hydroxypropanamido]butanoic acid; acetic acid
Synonyms: teriparatide
Molecular Weight (g/mol): 4177.83
Molecular Formula: C183H295N55O53S2
InChi Key: BUUKFBVDKSFMHN-LKMAISLMSA-N
SMILES: CC(O)=O.CC[C@H](C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@@H](N)CO)C(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC1=CN=CN1)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)