accessibility menu, dialog, popup

UserName

CAS RN 522-27-0

IUPAC Name: (E)-N-[(2Z)-1,2-bis(furan-2-yl)-2-(hydroxyimino)ethylidene]hydroxylamine
Synonyms: (E)-N-[(2Z)-1,2-bis(furan-2-yl)-2-(hydroxyimino)ethylidene]hydroxylamine
Molecular Weight (g/mol): 220.18
Molecular Formula: C10H8N2O4
InChi Key: RBOZTFPIXJBLPK-DSOJMZEYSA-N
SMILES: O\N=C(\C(=N/O)\C1=CC=CO1)/C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results