accessibility menu, dialog, popup

UserName

CAS RN 52127-83-0

IUPAC Name: 4-(2,6-dichloropyrimidin-4-yl)morpholine
Synonyms: 4-(2,6-dichloropyrimidin-4-yl)morpholine
Molecular Weight (g/mol): 234.08
Molecular Formula: C8H9Cl2N3O
InChi Key: QGGYMWHOBGSQCF-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=NC(=C1)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results