accessibility menu, dialog, popup

UserName

CAS RN 52093-25-1

IUPAC Name: europium(3+) tritrifluoromethanesulfonate
Synonyms: europium(3+) tritriflate
Molecular Weight (g/mol): 599.15
Molecular Formula: C3EuF9O9S3
InChi Key: TWNOVENTEPVGEJ-UHFFFAOYSA-K
SMILES: [Eu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results