accessibility menu, dialog, popup

UserName

CAS RN 51833-68-2

IUPAC Name: iron(3+) hydrate phosphate
Synonyms: iron(3+) hydrate phosphate
Molecular Weight (g/mol): 168.83
Molecular Formula: FeH2O5P
InChi Key: RQKNQWUJROKKEQ-UHFFFAOYSA-K
SMILES: O.[Fe+3].[O-]P([O-])([O-])=O
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)