accessibility menu, dialog, popup

UserName

CAS RN 518-18-3

IUPAC Name: 21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one
Synonyms: evodiamine
Molecular Weight (g/mol): 303.37
Molecular Formula: C19H17N3O
InChi Key: TXDUTHBFYKGSAH-UHFFFAOYNA-N
SMILES: CN1C2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results