accessibility menu, dialog, popup

UserName

CH3H3CCH3O(E)

CAS RN 5166-53-0

IUPAC Name: (3E)-5-methylhex-3-en-2-one
Synonyms: 3-hexen-2-one, 5-methyl-
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: IYMKNYVCXUEFJE-SNAWJCMRSA-N
SMILES: CC(C)\C=C\C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results