accessibility menu, dialog, popup

UserName

CAS RN 5158-50-9

IUPAC Name: bromodimethylborane
Synonyms: bromodimethylborane
Molecular Weight (g/mol): 120.78
Molecular Formula: C2H6BBr
InChi Key: ABQPEYRVNHDPIO-UHFFFAOYSA-N
SMILES: CB(C)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results