accessibility menu, dialog, popup

UserName

CAS RN 514-73-8

IUPAC Name: 3-ethyl-2-[(3E)-5-[(2E)-3-ethyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Synonyms: dithiazanine iodide
Molecular Weight (g/mol): 518.48
Molecular Formula: C23H23IN2S2
InChi Key: MNQDKWZEUULFPX-UHFFFAOYSA-M
SMILES: [I-].CCN1\C(SC2=CC=CC=C12)=C/C=C/C=CC1=[N+](CC)C2=CC=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results