CAS RN 51362-38-0
CAS RN 51362-38-0
IUPAC Name:
6-phenoxypyridine-3-carboxylic acid
Synonyms:
6-phenoxypyridine-3-carboxylic acid
Molecular Weight (g/mol):
215.21
Molecular Formula:
C12H9NO3
InChi Key:
GFEUNYLQJDQNAN-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CN=C(OC2=CC=CC=C2)C=C1
1
–
3
of
3
results
1
6-Phenoxynicotinic acid, 97%, Thermo Scientific™
CAS: 51362-38-0 Molecular Formula: C12H9NO3 Molecular Weight (g/mol): 215.208 MDL Number: MFCD02682068 InChI Key: GFEUNYLQJDQNAN-UHFFFAOYSA-N Synonym: 6-phenoxynicotinic acid,3-pyridinecarboxylic acid, 6-phenoxy,6-phenoxynicotinicacid,6-phenoxy-nicotinic acid,2-phenoxypyridine-5-carboxylic acid,3-pyridinecarboxylicacid, 6-phenoxy,6-phenyloxy-3-pyridinecarboxylic acid PubChem CID: 2776497 IUPAC Name: 6-phenoxypyridine-3-carboxylic acid SMILES: C1=CC=C(C=C1)OC2=NC=C(C=C2)C(=O)O