CAS RN 51322-75-9
IUPAC Name:
5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine
Synonyms:
tizanidine zanaflex sirdalud tizanidine hcl tizanidine hydrochloride ternelin tizanidina C9H8ClN5S tizanidina [inn-spanish] tizanidinum
Molecular Weight (g/mol):
253.71
Molecular Formula:
C9H8ClN5S
InChi Key:
XFYDIVBRZNQMJC-UHFFFAOYSA-N
SMILES:
ClC1=C(NC2=NCCN2)C2=NSN=C2C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Tizanidine hydrochloride, 99%
CAS: 51322-75-9 Molecular Formula: C9H8N5ClS.HCl