accessibility menu, dialog, popup

UserName

CAS RN 51312-42-6

IUPAC Name: Sodium dodecamolybdophosphate hydrate
Synonyms: —
Molecular Weight (g/mol): 2946.12
Molecular Formula: Na3O40PW12
InChi Key: —
SMILES: *
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)