accessibility menu, dialog, popup

UserName

CAS RN 51304-61-1

HClClNH

CAS RN 51304-61-1

IUPAC Name: hydrogen 4-(4-chlorophenyl)-1,2,3,6-tetrahydropyridine chloride
Synonyms: hydrogen 4-(4-chlorophenyl)-1,2,3,6-tetrahydropyridine chloride
Molecular Weight (g/mol): 230.13
Molecular Formula: C11H13Cl2N
InChi Key: NGDJUNABWNTULX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=C(C=C1)C1=CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)