accessibility menu, dialog, popup

UserName

CAS RN 51285-26-8

IUPAC Name: [amino(pyridin-2-yl)methylidene]azanium
Synonyms: [amino(pyridin-2-yl)methylidene]azanium
Molecular Weight (g/mol): 122.15
Molecular Formula: C6H8N3
InChi Key: KNXKVYCVGXFLES-UHFFFAOYSA-O
SMILES: NC(=[NH2+])C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results