CAS RN 51268-87-2
IUPAC Name:
3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol
Synonyms:
3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol
Molecular Weight (g/mol):
160.21
Molecular Formula:
C8H16O3
InChi Key:
BSLDYXOPGSOQGZ-ZETCQYMHSA-N
SMILES:
CC1(C)OC[C@H](CCCO)O1
1
–
1
of
1
results
1
Promotions Available
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]-propanol, 97%
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]-propanol, 97%
CAS: 51268-87-2 Molecular Formula: C8H16O3 Molecular Weight (g/mol): 160.21 MDL Number: MFCD01631673 InChI Key: BSLDYXOPGSOQGZ-ZETCQYMHSA-N Synonym: 3-4s-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol,1,3-dioxolane-4-propanol, 2,2-dimethyl-, s,s-3-2,2-dimethyl-1,3-dioxolan-4-yl propan-1-ol,3-4s-2,2-dimethyl-1,3-dioxolan-4-yl-propanol,1,3-dioxolane-4-propanol,2,2-dimethyl-, 4s,3-2,2-dimethyl-1,3-dioxolane-4 s-yl-propanol,3-2,2-dimethyl-1,3-dioxolane-4beta-yl-1-propanol PubChem CID: 2733890 IUPAC Name: 3-[(4S)-2,2-dimethyl-1,3-dioxolan-4-yl]propan-1-ol SMILES: CC1(OCC(O1)CCCO)C